siRNA Details
Gene Symbol: SLC1A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272087.1 | NP_001259016.1 | 8 | 2313 | ||
NM_001272088.1 | NP_001259017.1 | 6 | 1141 | |||
NM_005071.2 | NP_005062.1 | 5 | 919 | |||
NR_073589.1 | 5 | 872 | ||||
XM_006722842.1 | XP_006722905.1 | 6 | 1102 | |||
XM_006722844.1 | XP_006722907.1 | 6 | 995 | |||
XM_017027152.1 | XP_016882641.1 | 6 | 1190 | |||
GenBank mRNA | AK302380.1 | - | 5 | 849 | ||
AK312996.1 | - | 6 | 1185 | |||
BC028721.2 | - | 6 | 1063 | 5269300 | ||
BC040604.1 | - | 8 | 2268 | 5270965 | ||
U18244.1 | - | 5 | 919 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009200, D83262, AK159197, BC138140, BC138120.
Panther Classification:
Gene Ontology Categories:
