siRNA Details
| Gene Symbol: Cdca3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013538.5 | NP_038566.1 | 3 | 293 | ||
| GenBank mRNA | AK008606.1 | - | 3 | 246 | ||
| AK010409.1 | - | 3 | 234 | |||
| AK011313.1 | - | 3 | 266 | |||
| AK028159.1 | - | 3 | 235 | |||
| AK145537.1 | - | 3 | 235 | |||
| AK169792.1 | - | 3 | 241 | |||
| AK211930.1 | - | 2 | 194 | |||
| BC002006.1 | - | 3 | 247 | 3592108 | ||
| BC027172.1 | - | 3 | 214 | |||

Cdca3
Genomic Map