siRNA Details
| Gene Symbol: Hebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013546.2 | NP_038574.2 | 3 | 444 | ||
| GenBank mRNA | AA022130.1 | - | 432 | |||
| AB013095.1 | - | 3 | 383 | |||
| AF117613.1 | - | 3 | 432 | |||
| AK008919.1 | - | 3 | 365 | |||
| BC012654.1 | - | 3 | 368 | 4216585 | ||
| BY242687.1 | - | 3 | 445 | |||
| CT010262.1 | - | 3 | 326 | |||

Hebp1
Genomic Map