siRNA Details
| Gene Symbol: Hebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB013095.1 | - | 4 | 794 | ||
| BC012654.1 | - | 4 | 780 | 4216585 | ||
| CK378928.1 | - | 1 | 108 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hebp1| Gene Symbol: Hebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB013095.1 | - | 4 | 794 | ||
| BC012654.1 | - | 4 | 780 | 4216585 | ||
| CK378928.1 | - | 1 | 108 | |||