siRNA Details
| Gene Symbol: Hoxd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013554.5 | NP_038582.2 | 1, 2 | 844 | ||
| GenBank mRNA | AK134330.1 | - | 1, 2 | 846 | ||
| BC013463.1 | - | 1, 2 | 805 | 4240579 | ||
| BC048690.1 | - | 1, 2 | 787 | 6516538 | ||

Hoxd10
Genomic Map