siRNA Details
| Gene Symbol: Hprt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013556.2 | NP_038584.2 | 9 | 794 | ||
| GenBank mRNA | AK002286.1 | - | 9 | 739 | ||
| AK082827.1 | - | 9 | 683 | |||
| AK088114.1 | - | 9 | 684 | |||
| AK146626.1 | - | 9 | 740 | |||
| AY423851.1 | - | 3 | 149 | |||
| BC004686.1 | - | 9 | 709 | 3500885 | ||
| BC083145.1 | - | 9 | 680 | 30445781 | ||
| J00423.1 | - | 9 | 735 | |||

Hprt
Genomic Map