siRNA Details
| Gene Symbol: Htr3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099644.1 | NP_001093114.1 | 9 | 1778 | ||
| NM_013561.2 | NP_038589.2 | 9 | 1796 | |||
| GenBank mRNA | BC144799.1 | - | 9 | 1700 | 9053347 | |
| BC145743.1 | - | 9 | 1700 | 40131046 | ||
| CJ237116.1 | - | 1 | 218 | |||
| M74425.1 | - | 9 | 1747 | |||
| X72395.1 | - | 9 | 1662 | |||

Htr3a
Genomic Map