siRNA Details
| Gene Symbol: Ifrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013562.2 | NP_038590.2 | 11 | 1476 | ||
| GenBank mRNA | AK078888.1 | - | 1474 | |||
| AK168682.1 | - | 11 | 1472 | |||
| AK183550.1 | - | 3 | 227 | |||
| AK213011.1 | - | 2 | 123 | |||
| AK215747.1 | - | 1 | 46 | |||
| BC037013.1 | - | 11 | 1451 | |||
| BC043723.1 | - | 11 | 1443 | 5369100 | ||
| V00756.1 | - | 1 | 90 | |||
| X17400.1 | - | 11 | 1453 | |||

Ifrd1
Genomic Map