siRNA Details
| Gene Symbol: Incenp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016692.3 | NP_057901.2 | 4 | 526 | ||
| GenBank mRNA | AB100432.1 | - | 3 | 320 | ||
| AB100433.1 | - | 3 | 320 | |||
| AF117610.1 | - | 4 | 462 | |||
| AK034764.1 | - | 4 | 455 | |||
| AK081841.1 | - | 4 | 460 | |||
| AK088627.1 | - | 4 | 483 | |||
| BC021761.2 | - | 4 | 423 | |||
| BC037011.1 | - | 4 | 479 | |||
| BC052414.1 | - | 4 | 459 | 6401188 | ||

Incenp
Genomic Map