siRNA Details
| Gene Symbol: Incenp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016692.3 | NP_057901.2 | 17 | 2581 | ||
| GenBank mRNA | AB100432.1 | - | 16 | 2375 | ||
| AB100433.1 | - | 17 | 2387 | |||
| AF117610.1 | - | 17 | 2517 | |||
| AK034764.1 | - | 16 | 2841 | |||
| AK081841.1 | - | 17 | 2515 | |||
| AK088627.1 | - | 17 | 2538 | |||
| BC052414.1 | - | 17 | 2514 | 6401188 | ||

Incenp
Genomic Map