siRNA Details
Gene Symbol: Bcl11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242934.1 | NP_001229863.1 | 4 | 1060 | ||
NM_016707.3 | NP_057916.1 | 4 | 1060 | |||
GenBank mRNA | AF051525.1 | - | 4 | 998 | ||
AF186018.1 | - | 4 | 941 | |||
AK140949.1 | - | 4 | 1005 | |||
BC010585.1 | - | 1 | 74 | 2631265 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001191683, XM_008770430, XM_006251579, XM_006251591, XM_008770431, and others.
Panther Classification:
Gene Ontology Categories:
