siRNA Details
| Gene Symbol: Grk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018869.3 | NP_061357.3 | 14 | 2324 | ||
| GenBank mRNA | AF040746.1 | - | 14 | 1734 | ||
| AK135076.1 | - | 14 | 2326 | |||
| AK156465.1 | - | 13 | 2700 | |||
| AK161821.1 | - | 14 | 1804 | |||
| BC019379.1 | - | 14 | 1597 | 5116510 | ||

Grk5
Genomic Map