siRNA Details
| Gene Symbol: Grk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019497.2 | NP_062370.2 | 13 | 1917 | ||
| GenBank mRNA | AF040745.1 | - | 13 | 1343 | ||
| AK132685.1 | - | 13 | 1918 | |||
| BC150691.1 | - | 13 | 1441 | 9087602 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grk4| Gene Symbol: Grk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019497.2 | NP_062370.2 | 13 | 1917 | ||
| GenBank mRNA | AF040745.1 | - | 13 | 1343 | ||
| AK132685.1 | - | 13 | 1918 | |||
| BC150691.1 | - | 13 | 1441 | 9087602 | ||