siRNA Details
| Gene Symbol: FEZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042548.1 | NP_001036013.1 | 5 | 814 | ||
| NM_005102.2 | NP_005093.2 | 5 | 814 | |||
| XM_006712153.2 | XP_006712216.1 | 5 | 636 | |||
| GenBank mRNA | AB076184.1 | - | 5 | 806 | ||
| AB209719.1 | - | 5 | 613 | |||
| AF113124.1 | - | 5 | 713 | |||
| AK025274.1 | - | 4 | 494 | |||
| BC018032.1 | - | 5 | 793 | |||
| BC037566.1 | - | 5 | 779 | |||
| BC047020.1 | - | 5 | 765 | |||
| BC054043.1 | - | 5 | 748 | |||
| BC064484.1 | - | 5 | 728 | |||
| BC089390.1 | - | 5 | 760 | |||
| BC110386.1 | - | 5 | 769 | 5183735 | ||
| BX537785.1 | - | 6 | 740 | |||
| U60061.1 | - | 2 | 167 | |||
| U69140.1 | - | 4 | 455 | |||

FEZ2
Genomic Map