siRNA Details
| Gene Symbol: Epha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023580.4 | NP_076069.2 | 3 | 339 | ||
| GenBank mRNA | AF131197.1 | - | 3 | 281 | ||
| AK017662.1 | - | 3 | 307 | |||
| AK028478.1 | - | 3 | 340 | |||
| BC071215.1 | - | 3 | 303 | 4196138 | ||

Epha1
Genomic Map