siRNA Details
| Gene Symbol: Epha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023580.4 | NP_076069.2 | 4 | 796 | ||
| GenBank mRNA | AF131197.1 | - | 4 | 738 | ||
| AK028478.1 | - | 4 | 797 | |||
| BC071215.1 | - | 4 | 760 | 4196138 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epha1| Gene Symbol: Epha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023580.4 | NP_076069.2 | 4 | 796 | ||
| GenBank mRNA | AF131197.1 | - | 4 | 738 | ||
| AK028478.1 | - | 4 | 797 | |||
| BC071215.1 | - | 4 | 760 | 4196138 | ||