siRNA Details
| Gene Symbol: Amz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252193.1 | NP_001239122.1 | 3 | 622 | ||
| NM_025275.4 | NP_079551.3 | 4 | 441 | |||
| GenBank mRNA | AJ879914.1 | - | 1 | 138 | ||
| AK075748.1 | - | 4 | 381 | |||
| AK143908.1 | - | 4 | 406 | |||
| AK182279.1 | - | 2 | 235 | |||
| BC025087.1 | - | 3 | 616 | 2649126 | ||

Amz2
Genomic Map