siRNA Details
| Gene Symbol: Ifrd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025903.2 | NP_080179.1 | 12 | 1921 | ||
| GenBank mRNA | AK002702.1 | - | 6 | 1146 | ||
| AK007687.1 | - | 12 | 1922 | |||
| AK149496.1 | - | 12 | 1902 | |||
| AK156625.1 | - | 11 | 1997 | |||
| AK165967.1 | - | 12 | 1914 | |||
| AK166130.1 | - | 12 | 1989 | |||
| AK166258.1 | - | 12 | 1879 | |||
| BC023486.1 | - | 12 | 1868 | |||
| BC037434.1 | - | 12 | 1877 | 2651172 | ||

Ifrd2
Genomic Map