siRNA Details
Gene Symbol: Id4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031166.2 | NP_112443.1 | 3 | 798 | ||
GenBank mRNA | X75018.1 | - | 3 | 798 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Id4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031166.2 | NP_112443.1 | 3 | 798 | ||
GenBank mRNA | X75018.1 | - | 3 | 798 |