siRNA Details
| Gene Symbol: Id4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031166.2 | NP_112443.1 | 3 | 801 | ||
| GenBank mRNA | X75018.1 | - | 3 | 801 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Id4| Gene Symbol: Id4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031166.2 | NP_112443.1 | 3 | 801 | ||
| GenBank mRNA | X75018.1 | - | 3 | 801 | ||