siRNA Details
| Gene Symbol: Ftcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080845.2 | NP_543121.1 | 7 | 913 | ||
| GenBank mRNA | AK165414.1 | - | 6 | 822 | ||
| BC010813.1 | - | 7 | 875 | 4211065 | ||
| BC024078.1 | - | 7 | 907 | 5098529 | ||
| CB949427.1 | - | 1 | 51 | |||

Ftcd
Genomic Map