siRNA Details
| Gene Symbol: Il13ra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080147.1 | - | 2 | 134 | ||
| AK082889.1 | - | 2 | 129 | |||
| AK089984.1 | - | 2 | 165 | |||
| AK143292.1 | - | 2 | 134 | |||
| AK154675.1 | - | 2 | 189 | |||
| BC052425.1 | - | 2 | 157 | 6835500 | ||
| BC059939.1 | - | 2 | 125 | 6477224 | ||
| S80963.1 | - | 2 | 162 | |||

Il13ra1
Genomic Map