siRNA Details
Gene Symbol: CCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005125.1 | NP_005116.1 | 7 | 682 | ||
GenBank mRNA | AF002210.1 | - | 7 | 682 | ||
AY113179.1 | - | 7 | 702 | |||
BC105016.1 | - | 7 | 678 | 8144019 | ||
BC112055.1 | - | 7 | 678 | 8327523 | ||
CR541928.1 | - | 7 | 639 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
