siRNA Details
| Gene Symbol: Jchain | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152839.3 | NP_690052.2 | 4 | 658 | ||
| GenBank mRNA | AK078865.1 | - | 4 | 544 | ||
| AK160344.1 | - | 4 | 610 | |||
| BC006026.1 | - | 4 | 562 | 3491646 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jchain| Gene Symbol: Jchain | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152839.3 | NP_690052.2 | 4 | 658 | ||
| GenBank mRNA | AK078865.1 | - | 4 | 544 | ||
| AK160344.1 | - | 4 | 610 | |||
| BC006026.1 | - | 4 | 562 | 3491646 | ||