siRNA Details
| Gene Symbol: Hoxc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175730.5 | NP_783857.1 | 2 | 735 | ||
| GenBank mRNA | AK078527.1 | - | 2 | 736 | ||
| BC115551.1 | - | 2 | 736 | 40101712 | ||
| BC115552.1 | - | 2 | 736 | 40101713 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoxc5| Gene Symbol: Hoxc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175730.5 | NP_783857.1 | 2 | 735 | ||
| GenBank mRNA | AK078527.1 | - | 2 | 736 | ||
| BC115551.1 | - | 2 | 736 | 40101712 | ||
| BC115552.1 | - | 2 | 736 | 40101713 | ||