siRNA Details
| Gene Symbol: Hs3st3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178870.5 | NP_849201.1 | 2 | 2071 | ||
| GenBank mRNA | AK032282.1 | - | 2 | 1039 | ||
| AK051284.1 | - | 2 | 2072 | |||
| BC094320.1 | - | 2 | 2125 | 6400013 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hs3st3a1| Gene Symbol: Hs3st3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178870.5 | NP_849201.1 | 2 | 2071 | ||
| GenBank mRNA | AK032282.1 | - | 2 | 1039 | ||
| AK051284.1 | - | 2 | 2072 | |||
| BC094320.1 | - | 2 | 2125 | 6400013 | ||