siRNA Details
| Gene Symbol: Api5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK079865.1 | - | 2 | 257 | ||
| AK134480.1 | - | 2 | 310 | |||
| AK137466.1 | - | 2 | 339 | |||
| AK153934.1 | - | 2 | 283 | |||
| AK160874.1 | - | 2 | 307 | |||
| AK163586.1 | - | 2 | 314 | |||
| AK167315.1 | - | 2 | 283 | |||
| AK167375.1 | - | 2 | 323 | |||
| AK178132.1 | - | 2 | 200 | |||
| BC007133.1 | - | 2 | 302 | 3594358 | ||
| U35846.1 | - | 2 | 267 | |||

Api5
Genomic Map