siRNA Details
| Gene Symbol: Atp5k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007507.2 | NP_031533.2 | 4 | 228 | ||
| GenBank mRNA | AK011342.1 | - | 3 | 138 | ||
| BC028438.1 | - | 4 | 228 | 1332765 | ||
| BC089550.1 | - | 4 | 220 | 30380642 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp5k| Gene Symbol: Atp5k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007507.2 | NP_031533.2 | 4 | 228 | ||
| GenBank mRNA | AK011342.1 | - | 3 | 138 | ||
| BC028438.1 | - | 4 | 228 | 1332765 | ||
| BC089550.1 | - | 4 | 220 | 30380642 | ||