siRNA Details
| Gene Symbol: Bach2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109661.1 | NP_001103131.1 | 3 | 463 | ||
| GenBank mRNA | BC131642.1 | - | 3 | 371 | 40109815 | |
| CX200493.1 | - | 1 | 16 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bach2| Gene Symbol: Bach2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109661.1 | NP_001103131.1 | 3 | 463 | ||
| GenBank mRNA | BC131642.1 | - | 3 | 371 | 40109815 | |
| CX200493.1 | - | 1 | 16 | |||