siRNA Details
| Gene Symbol: Blk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007549.2 | NP_031575.2 | 9 | 1234 | ||
| GenBank mRNA | AK089888.1 | - | 9 | 1234 | ||
| AK171640.1 | - | 9 | 1181 | |||
| BC030668.1 | - | 9 | 1084 | 1178763 | ||
| CT010240.1 | - | 8 | 814 | |||
| M30903.1 | - | 9 | 1163 | |||

Blk
Genomic Map