siRNA Details
| Gene Symbol: Bmpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007561.4 | NP_031587.1 | 11, 12 | 1967 | ||
| GenBank mRNA | AF003942.1 | - | 11, 12 | 1967 | ||
| AK138715.1 | - | 1, 2 | 95 | |||
| AK148180.1 | - | 6, 7 | 785 | |||
| AK149385.1 | - | 2848 | ||||
| U78048.1 | - | 11, 12 | 1609 | |||

Bmpr2
Genomic Map