siRNA Details
| Gene Symbol: C1qc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007574.2 | NP_031600.2 | 3 | 434 | ||
| GenBank mRNA | AK138974.1 | - | 436 | |||
| BB636871.1 | - | 3 | 435 | |||
| BC043945.1 | - | 3 | 414 | 4972540 | ||
| BC054443.1 | - | 3 | 405 | 6475447 | ||
| BC075724.1 | - | 3 | 413 | 3675763 | ||
| CK332232.1 | - | 3 | 395 | |||
| X66295.1 | - | 3 | 400 | |||

C1qc
Genomic Map