siRNA Details
| Gene Symbol: Ciita | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF042158.1 | - | 19 | 4023 | ||
| AF042159.1 | - | 20 | 4329 | |||
| AF100709.1 | - | 19 | 4206 | |||
| AF100710.1 | - | 19 | 3922 | |||
| AK138822.1 | - | 19 | 4112 | |||
| AK153609.1 | - | 19 | 4019 | |||
| AK155184.1 | - | 19 | 4046 | |||
| U60653.1 | - | 19 | 3983 | |||

Ciita
Genomic Map