siRNA Details
| Gene Symbol: Cacng2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007583.2 | NP_031609.1 | 2 | 620 | ||
| GenBank mRNA | AF077739.1 | - | 2 | 620 | ||
| AK045362.1 | - | 1369 | ||||
| BC103563.1 | - | 2 | 301 | 40045028 | ||
| BC103564.1 | - | 2 | 300 | 40045029 | ||
| BC103664.1 | - | 2 | 300 | 40045030 | ||

Cacng2
Genomic Map