siRNA Details
| Gene Symbol: Cbx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001076789.1 | NP_001070257.1 | 5 | 2091 | ||
| NM_001110216.1 | NP_001103686.1 | 6 | 2192 | |||
| NM_007626.3 | NP_031652.1 | 5 | 2092 | |||
| GenBank mRNA | AK008792.1 | - | 5 | 2127 | ||
| AK011031.1 | - | 1 | 840 | |||
| AK030366.1 | - | 5 | 2092 | |||
| AK032975.1 | - | 5 | 2048 | |||
| BC004707.1 | - | 5 | 2091 | 3499489 | ||

Cbx5
Genomic Map