siRNA Details
Gene Symbol: Ccnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007630.2 | NP_031656.2 | 6 | 847 | ||
GenBank mRNA | AK013312.1 | - | 6 | 816 | ||
AK076122.1 | - | 6 | 841 | |||
AK177442.1 | - | 1 | 119 | |||
BC008247.1 | - | 6 | 763 | 3156950 | ||
CT010286.1 | - | 6 | 733 | |||
X66032.1 | - | 6 | 834 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
