siRNA Details
| Gene Symbol: Ccne1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007633.2 | NP_031659.2 | 3, 4 | 293 | ||
| GenBank mRNA | AK089950.1 | - | 3, 4 | 225 | ||
| AK163046.1 | - | 3, 4 | 282 | |||
| AK165658.1 | - | 3, 4 | 283 | |||
| BC062152.1 | - | 3, 4 | 320 | 6510163 | ||
| BC084588.1 | - | 3, 4 | 337 | 6506897 | ||
| BC106191.1 | - | 3, 4 | 293 | 6336006 | ||
| BC138660.1 | - | 2, 3 | 126 | 8861680 | ||
| BC138662.1 | - | 2, 3 | 126 | 8861683 | ||
| X75888.1 | - | 2, 3 | 133 | |||

Ccne1
Genomic Map