siRNA Details
| Gene Symbol: Scarb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007644.3 | NP_031670.1 | 8 | 1388 | ||
| GenBank mRNA | AB008553.1 | - | 8 | 1293 | ||
| AK011123.1 | - | 8 | 1318 | |||
| AK083038.1 | - | 8 | 1389 | |||
| AK144235.1 | - | 8 | 1420 | |||
| AK170555.1 | - | 48 | ||||
| AK185315.1 | - | 2 | 231 | |||
| BC029073.1 | - | 8 | 1296 | 3962669 | ||

Scarb2
Genomic Map