siRNA Details
| Gene Symbol: Cd3e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007648.4 | NP_031674.1 | 5 | 227 | ||
| GenBank mRNA | BC145926.1 | - | 5 | 227 | 40131229 | |
| BC145954.1 | - | 5 | 227 | 40131257 | ||
| EF474454.2 | - | 4 | 149 | |||
| GU456631.1 | - | 4 | 149 | |||
| J02990.1 | - | 5 | 228 | |||

Cd3e
Genomic Map