siRNA Details
| Gene Symbol: Cd53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007651.3 | NP_031677.1 | 5 | 606 | ||
| GenBank mRNA | AA177409.1 | - | 5 | 494 | ||
| AK156510.1 | - | 5 | 513 | |||
| AK160326.1 | - | 5 | 504 | |||
| BC021310.1 | - | 5 | 520 | 5010267 | ||
| BC052905.1 | - | 5 | 471 | 30078163 | ||
| X97227.1 | - | 4 | 407 | |||
| Z16071.1 | - | 4 | 392 | |||

Cd53
Genomic Map