siRNA Details
| Gene Symbol: Cdk11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007661.3 | NP_031687.2 | 4 | 361 | ||
| GenBank mRNA | AK008283.1 | - | 4 | 338 | ||
| AK077668.1 | - | 4 | 362 | |||
| AK135402.1 | - | 4 | 368 | |||
| AK147133.1 | - | 4 | 339 | |||
| BC052920.1 | - | 4 | 439 | 6489832 | ||
| L37092.1 | - | 3 | 936 | |||

Cdk11b
Genomic Map