siRNA Details
| Gene Symbol: Cdx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007673.3 | NP_031699.2 | 3 | 1440 | ||
| GenBank mRNA | AK018629.1 | - | 1 | 464 | ||
| AK049566.1 | - | 3 | 1441 | |||
| AK049574.1 | - | 3 | 1313 | |||
| BC103516.1 | - | 3 | 1318 | 40039012 | ||
| BC103517.1 | - | 3 | 1321 | 40039015 | ||
| BC103519.1 | - | 3 | 1321 | 40039019 | ||
| BC103520.1 | - | 3 | 1321 | 40039017 | ||

Cdx2
Genomic Map