siRNA Details
| Gene Symbol: HIST1H1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005319.3 | NP_005310.1 | 1 | 408 | ||
| GenBank mRNA | AB451259.1 | - | 1 | 365 | ||
| AB451385.1 | - | 1 | 365 | |||
| AK290947.1 | - | 1 | 407 | |||
| BC002649.1 | - | 1 | 409 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIST1H1C| Gene Symbol: HIST1H1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005319.3 | NP_005310.1 | 1 | 408 | ||
| GenBank mRNA | AB451259.1 | - | 1 | 365 | ||
| AB451385.1 | - | 1 | 365 | |||
| AK290947.1 | - | 1 | 407 | |||
| BC002649.1 | - | 1 | 409 | |||