siRNA Details
| Gene Symbol: Chrm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001112697.1 | NP_001106167.1 | 3 | 3608 | ||
| NM_007698.3 | NP_031724.2 | 2 | 3516 | |||
| GenBank mRNA | AK081248.1 | - | 2 | 3173 | ||
More Information
Set Membership: |
|

Chrm1
Genomic Map