siRNA Details
| Gene Symbol: HIST1H1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005321.2 | NP_005312.1 | 1 | 638 | ||
| GenBank mRNA | AK313681.1 | - | 1 | 639 | ||
| BC096168.1 | - | 1 | 610 | 40003408 | ||
| BC096169.1 | - | 1 | 610 | 40003411 | ||
| BC096170.1 | - | 1 | 610 | 40003413 | ||
| BC099632.1 | - | 1 | 610 | 40003410 | ||
| EF065554.1 | - | 1 | 606 | |||
| EF065555.1 | - | 1 | 606 | |||

HIST1H1E
Genomic Map