siRNA Details
| Gene Symbol: Clock | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289826.1 | NP_001276755.1 | 6 | 489 | ||
| NM_007715.6 | NP_031741.1 | 6 | 532 | |||
| GenBank mRNA | AB221635.1 | - | 3 | 144 | ||
| AF000998.1 | - | 6 | 532 | |||
| AK030352.1 | - | 6 | 454 | |||
| AK040337.1 | - | 6 | 470 | |||
| AK041303.1 | - | 6 | 470 | |||
| AK129118.1 | - | 6 | 489 | |||
| AK158765.1 | - | 6 | 470 | |||
| BC129886.1 | - | 4 | 241 | |||

Clock
Genomic Map