siRNA Details
| Gene Symbol: Clock | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289826.1 | NP_001276755.1 | 13, 14 | 1331 | ||
| NM_007715.6 | NP_031741.1 | 13, 14 | 1374 | |||
| GenBank mRNA | AB221635.1 | - | 10, 11 | 986 | ||
| AF000998.1 | - | 13, 14 | 1374 | |||
| AK030352.1 | - | 13, 14 | 1274 | |||
| AK129118.1 | - | 13, 14 | 1331 | |||
| BC129886.1 | - | 11, 12 | 1082 | |||

Clock
Genomic Map