siRNA Details
| Gene Symbol: Ackr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271607.1 | NP_001258536.1 | 3 | 532 | ||
| NM_007722.4 | NP_031748.2 | 2 | 249 | |||
| GenBank mRNA | AF000236.1 | - | 2 | 203 | ||
| AK031100.1 | - | 227 | ||||
| BC015254.1 | - | 2 | 164 | 4242244 | ||

Ackr3
Genomic Map