siRNA Details
| Gene Symbol: Ackr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271607.1 | NP_001258536.1 | 3 | 2005 | ||
| NM_007722.4 | NP_031748.2 | 2 | 1722 | |||
| GenBank mRNA | AF000236.1 | - | 2 | 1662 | ||
| AK031100.1 | - | 1700 | ||||
| BC015254.1 | - | 2 | 1623 | 4242244 | ||
More Information
Other Species Targeted
Set Membership: |
|

Ackr3
Genomic Map