siRNA Details
| Gene Symbol: Cnr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007726.3 | NP_031752.1 | 2 | 2146 | ||
| GenBank mRNA | BC070447.1 | - | 2 | 2146 | 30619104 | |
| BC075644.1 | - | 6 | 2563 | |||
| BC079564.1 | - | 1 | 1630 | 30536039 | ||
| CD351242.1 | - | 1 | 524 | |||
More Information
Set Membership: |
|

Cnr1
Genomic Map